Product Details
- SNP ID
-
rs72718362
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:140623477 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGAGTCAAGAAGGACTATGTCATA[C/T]ATTTGGCTCAGTAACTTTTTTCTTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TBC1D9
PubMed Links
Gene Details
- Gene
- TBC1D9
- Gene Name
- TBC1 domain family member 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015130.2 |
|
Intron |
|
|
NP_055945.2 |
View Full Product Details