Product Details

SNP ID
rs72931491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:83462358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTATTACTGCAAACAGGTGAACA[C/T]AGTAAAAACAAATGAACTTACTGCA
Phenotype
MIM: 611143 MIM: 606769 MIM: 611983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM175A PubMed Links

Gene Details

Gene
FAM175A
Gene Name
family with sequence similarity 175 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139076.2 1312 Intron NP_620775.2
XM_005263280.4 1312 UTR 3 XP_005263337.1
XM_017008694.1 1312 UTR 3 XP_016864183.1
Gene
HELQ
Gene Name
helicase, POLQ-like
There are no transcripts associated with this gene.

Gene
MRPS18C
Gene Name
mitochondrial ribosomal protein S18C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297767.1 1312 Intron NP_001284696.1
NM_001297769.1 1312 Intron NP_001284698.1
NM_001297770.1 1312 Intron NP_001284699.1
NM_016067.3 1312 Intron NP_057151.1

View Full Product Details