Product Details

SNP ID
rs72987021
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:109555517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCACTTTTGCAGTCTGAAAATA[C/T]GGCCCTTTTCAGGAATTAAAATAAT
Phenotype
MIM: 139370 MIM: 606916
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNAI3 PubMed Links

Gene Details

Gene
GNAI3
Gene Name
G protein subunit alpha i3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006496.3 Intron NP_006487.1
Gene
GPR61
Gene Name
G protein-coupled receptor 61
There are no transcripts associated with this gene.

View Full Product Details