Product Details

SNP ID
rs73142320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98737715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATATCACCAGCATTTTATCTTG[C/T]CGTTTTAGTTTGGCCTTTTTCTGTG
Phenotype
MIM: 607815 MIM: 602233
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 Intron NP_001190994.1
NM_001204066.1 Intron NP_001190995.1
NM_001204067.1 Intron NP_001190996.1
NM_001204068.1 Intron NP_001190997.1
NM_001204069.1 Intron NP_001190998.1
NM_001204070.1 Intron NP_001190999.1
NM_001204079.1 Intron NP_001191008.1
NM_001204080.1 Intron NP_001191009.1
NM_001204081.1 Intron NP_001191010.1
NM_020140.3 Intron NP_064525.1
NM_152788.4 Intron NP_690001.3
NM_181670.3 Intron NP_858056.2
XM_005269028.4 Intron XP_005269085.1
XM_005269029.4 Intron XP_005269086.1
XM_005269032.3 Intron XP_005269089.1
XM_006719504.3 Intron XP_006719567.1
XM_006719505.3 Intron XP_006719568.1
XM_006719506.3 Intron XP_006719569.1
XM_006719507.3 Intron XP_006719570.1
XM_006719508.3 Intron XP_006719571.1
XM_006719509.3 Intron XP_006719572.1
XM_006719510.3 Intron XP_006719573.1
XM_006719512.3 Intron XP_006719575.1
XM_006719513.3 Intron XP_006719576.1
XM_006719514.3 Intron XP_006719577.1
XM_011538571.2 Intron XP_011536873.1
XM_017019651.1 Intron XP_016875140.1
XM_017019652.1 Intron XP_016875141.1
XM_017019653.1 Intron XP_016875142.1
XM_017019654.1 Intron XP_016875143.1
XM_017019655.1 Intron XP_016875144.1
XM_017019656.1 Intron XP_016875145.1
XM_017019657.1 Intron XP_016875146.1
XM_017019658.1 Intron XP_016875147.1
XM_017019659.1 Intron XP_016875148.1
XM_017019660.1 Intron XP_016875149.1
XM_017019661.1 Intron XP_016875150.1
XM_017019662.1 Intron XP_016875151.1
XM_017019663.1 Intron XP_016875152.1
XM_017019664.1 Intron XP_016875153.1
XM_017019665.1 Intron XP_016875154.1
Gene
APAF1
Gene Name
apoptotic peptidase activating factor 1
There are no transcripts associated with this gene.

View Full Product Details