Product Details

SNP ID
rs73120283
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:223221620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTTTCGTCTTGCTTCGAGTAG[A/C]GTTGTCTGGATGAAGGAAGATTCTC
Phenotype
MIM: 615827
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SUSD4 PubMed Links

Gene Details

Gene
SUSD4
Gene Name
sushi domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037175.2 2264 Intron NP_001032252.1
NM_017982.3 2264 UTR 3 NP_060452.3
XM_005273169.1 2264 UTR 3 XP_005273226.1
XM_005273172.1 2264 UTR 3 XP_005273229.1
XM_006711408.1 2264 UTR 3 XP_006711471.1
XM_011509685.1 2264 UTR 3 XP_011507987.1
XM_011509687.1 2264 UTR 3 XP_011507989.1
XM_017001583.1 2264 UTR 3 XP_016857072.1
XM_017001584.1 2264 UTR 3 XP_016857073.1
XM_017001585.1 2264 UTR 3 XP_016857074.1
XM_017001586.1 2264 UTR 3 XP_016857075.1
XM_017001587.1 2264 UTR 3 XP_016857076.1
XM_017001588.1 2264 UTR 3 XP_016857077.1
XM_017001589.1 2264 Intron XP_016857078.1

View Full Product Details