Product Details

SNP ID
rs73093819
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:209831030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGAGATGGCTGCAGAGTCTAC[C/T]GAAAGTCCAGAGAGTAAGTGTCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIEXF PubMed Links

Gene Details

Gene
DIEXF
Gene Name
digestive organ expansion factor homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014388.6 472 Silent Mutation ACC,ACT T125T NP_055203.4
XM_006711275.3 472 Silent Mutation ACC,ACT T125T XP_006711338.1
XM_011509418.2 472 UTR 5 XP_011507720.1

View Full Product Details