Product Details

SNP ID
rs72832011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68184053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAATATGCTTACTGCCACTTAGC[C/T]CCCCAACACATATACCTCATCTCTG
Phenotype
MIM: 601302
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PNO1 PubMed Links

Gene Details

Gene
PNO1
Gene Name
partner of NOB1 homolog
There are no transcripts associated with this gene.

Gene
PPP3R1
Gene Name
protein phosphatase 3 regulatory subunit B, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000945.3 Intron NP_000936.1

View Full Product Details