Product Details

SNP ID
rs73106063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:4979826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTGGCACTCCTTGGCCCTTCAA[C/G]CCTCAACTGCAGCCCCATGTTATGC
Phenotype
MIM: 604256
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BHLHE40 PubMed Links

Gene Details

Gene
BHLHE40
Gene Name
basic helix-loop-helix family member e40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003670.2 Intron NP_003661.1
Gene
BHLHE40-AS1
Gene Name
BHLHE40 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details