Product Details

SNP ID
rs68003332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:59751950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAACATTGGATTCTGGAGGAGCA[A/C]CACCGCAGGAAAGACTGGAGAAAGG
Phenotype
MIM: 601153
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FHIT PubMed Links

Gene Details

Gene
FHIT
Gene Name
fragile histidine triad
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166243.2 1808 Intron NP_001159715.1
NM_001320899.1 1808 UTR 3 NP_001307828.1
NM_001320900.1 1808 UTR 3 NP_001307829.1
NM_001320901.1 1808 UTR 3 NP_001307830.1
NM_002012.3 1808 Intron NP_002003.1
XM_017005880.1 1808 UTR 3 XP_016861369.1
XM_017005881.1 1808 UTR 3 XP_016861370.1
XM_017005882.1 1808 UTR 3 XP_016861371.1
XM_017005883.1 1808 Intron XP_016861372.1
XM_017005884.1 1808 Intron XP_016861373.1
XM_017005885.1 1808 UTR 3 XP_016861374.1
XM_017005886.1 1808 UTR 3 XP_016861375.1

View Full Product Details