Product Details

SNP ID
rs71296764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:62371471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAATCTTTGGCCATCGTATCCCC[A/G]GTGTTATCACTAAGATTCTGGGGGT
Phenotype
MIM: 607414
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FEZF2 PubMed Links

Gene Details

Gene
FEZF2
Gene Name
FEZ family zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018008.3 Intron NP_060478.3

View Full Product Details