Product Details
- SNP ID
-
rs72702229
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:174241074 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAAACATTACTTAACTCATTTTG[A/G]ATGAAAAGTTAATTTTCGTTTTTAC
- Phenotype
-
MIM: 605649
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO8
PubMed Links
Gene Details
- Gene
- FBXO8
- Gene Name
- F-box protein 8
View Full Product Details