Product Details

SNP ID
rs72825454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:166981472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCAATCCTAAGAACCCAAGGTA[C/T]CTAAAGCGTCAGGTTCCCTCCTTCT
Phenotype
MIM: 610119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TENM2 PubMed Links

Gene Details

Gene
TENM2
Gene Name
teneurin transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080428.2 Intron NP_001073897.2
NM_001122679.1 Intron NP_001116151.1
XM_005265950.2 Intron XP_005266007.1
XM_005265952.2 Intron XP_005266009.1
XM_006714897.2 Intron XP_006714960.1
XM_011534604.2 Intron XP_011532906.1
XM_017009660.1 Intron XP_016865149.1
XM_017009661.1 Intron XP_016865150.1
XM_017009662.1 Intron XP_016865151.1
XM_017009663.1 Intron XP_016865152.1
XM_017009664.1 Intron XP_016865153.1
XM_017009665.1 Intron XP_016865154.1
XM_017009666.1 Intron XP_016865155.1
XM_017009667.1 Intron XP_016865156.1
XM_017009668.1 Intron XP_016865157.1
XM_017009669.1 Intron XP_016865158.1

View Full Product Details