Product Details

SNP ID
rs73801493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:171864911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAACCCACTCAGATACTGTCACA[A/G]TAAGTGATGCACTAAACCACTGGCC
Phenotype
MIM: 605651
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXW11 PubMed Links

Gene Details

Gene
FBXW11
Gene Name
F-box and WD repeat domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012300.2 Intron NP_036432.2
NM_033644.2 Intron NP_387448.2
NM_033645.2 Intron NP_387449.2
XM_005265855.4 Intron XP_005265912.1
XM_005265856.2 Intron XP_005265913.1
XM_005265857.2 Intron XP_005265914.1
XM_005265858.2 Intron XP_005265915.1
XM_017009279.1 Intron XP_016864768.1
XM_017009280.1 Intron XP_016864769.1
XM_017009281.1 Intron XP_016864770.1

View Full Product Details