Product Details

SNP ID
rs72554027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173233110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCATAGACCTGCGCCTGCGAG[G/A]AGAGCACGCGCGGCTTCCTCCGCCG
Phenotype
MIM: 600584
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
NKX2-5 PubMed Links

Gene Details

Gene
NKX2-5
Gene Name
NK2 homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166175.1 918 UTR 3 NP_001159647.1
NM_001166176.1 918 UTR 3 NP_001159648.1
NM_004387.3 918 Missense Mutation TCC,TTC S145F NP_004378.1
XM_017009071.1 918 Intron XP_016864560.1

View Full Product Details