Product Details

SNP ID
rs72825319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176320784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAAATAGGATTAAATAGGATAG[C/T]TCTTCTCTGCTGCTGGCTTAGGATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIMC1 PubMed Links

Gene Details

Gene
SIMC1
Gene Name
SUMO interacting motifs containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308195.1 Intron NP_001295124.1
NM_001308196.1 Intron NP_001295125.1
NM_001308200.1 Intron NP_001295129.1
NM_198567.5 Intron NP_940969.3
XM_011534553.2 Intron XP_011532855.1
XM_011534554.2 Intron XP_011532856.1
XM_011534556.2 Intron XP_011532858.1
XM_017009454.1 Intron XP_016864943.1
XM_017009455.1 Intron XP_016864944.1
XM_017009456.1 Intron XP_016864945.1

View Full Product Details