Product Details

SNP ID
rs73811151
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179813771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTCTCAAAATAAAATAAAATA[A/T]AACCTTTTGGGCAGGCATGGAAAGG
Phenotype
MIM: 604561 MIM: 601530
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
MGAT4B PubMed Links
Additional Information
For this assay, SNP(s) [rs201565764] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MGAT4B
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
There are no transcripts associated with this gene.

Gene
SQSTM1
Gene Name
sequestosome 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142298.1 1988 Intron NP_001135770.1
NM_001142299.1 1988 Intron NP_001135771.1
NM_003900.4 1988 Intron NP_003891.1
XM_011534683.2 1988 UTR 3 XP_011532985.1
XM_017010010.1 1988 Intron XP_016865499.1

View Full Product Details