Product Details
- SNP ID
-
rs71599703
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:163505747 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTCCCGGGAGCTGTCGGCTGGTG[A/G]AGGTGAGGGAGTCGGCCTGGGCGTC
- Phenotype
-
MIM: 605527
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MAT2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs185678887,rs369444943] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAT2B
- Gene Name
- methionine adenosyltransferase 2B
View Full Product Details