Product Details

SNP ID
rs73715685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143567831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGCAGGGGGTCCTGTGGACTCC[A/G]CAGGTAGCCCTGGGTCTGGCTCAGG
Phenotype
MIM: 611552
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSDMD PubMed Links

Gene Details

Gene
GSDMD
Gene Name
gasdermin D
There are no transcripts associated with this gene.

Gene
MROH6
Gene Name
maestro heat like repeat family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100878.1 2153 Missense Mutation CGG,TGG R608W NP_001094348.1
XM_006716615.2 2153 Intron XP_006716678.1
XM_011517214.1 2153 Missense Mutation GCG,GTG A693V XP_011515516.1
XM_011517215.1 2153 Missense Mutation GCG,GTG A692V XP_011515517.1
XM_011517216.2 2153 Missense Mutation GCG,GTG A693V XP_011515518.1
XM_011517217.1 2153 Missense Mutation GCG,GTG A693V XP_011515519.1
XM_011517218.1 2153 Missense Mutation CGG,TGG R569W XP_011515520.1
XM_011517219.1 2153 UTR 3 XP_011515521.1
XM_011517220.1 2153 UTR 3 XP_011515522.1
XM_011517221.2 2153 Missense Mutation CGG,TGG R615W XP_011515523.1
XM_011517222.1 2153 Missense Mutation CGG,TGG R615W XP_011515524.1
XM_011517223.1 2153 Missense Mutation CGG,TGG R615W XP_011515525.1
Gene
NAPRT
Gene Name
nicotinate phosphoribosyltransferase
There are no transcripts associated with this gene.

View Full Product Details