Product Details

SNP ID
rs111633101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87882189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCACACATGTTCCAAAACTTAGG[A/G]AAGTGAGGACTGTATTTGGGGCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC497256 PubMed Links

Gene Details

Gene
LOC497256
Gene Name
uncharacterized LOC497256
There are no transcripts associated with this gene.

Gene
SPATA31E1
Gene Name
SPATA31 subfamily E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178828.4 Intron NP_849150.3

View Full Product Details