Product Details

SNP ID
rs73162469
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133132869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGAAGTTTACAGTGAATACTTA[C/T]AAACCTTCCCATCTAGACTCTACCA
Phenotype
MIM: 194538
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF10 PubMed Links

Gene Details

Gene
ZNF10
Gene Name
zinc finger protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015394.4 Intron NP_056209.2
Gene
ZNF891
Gene Name
zinc finger protein 891
There are no transcripts associated with this gene.

View Full Product Details