Product Details

SNP ID
rs73181129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41463755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAATATTCTGGATCTACGGCT[C/T]ATTCTCTACAGATTCTCTCCAGGCC
Phenotype
MIM: 610077
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGCC PubMed Links

Gene Details

Gene
RGCC
Gene Name
regulator of cell cycle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014059.2 Intron NP_054778.2

View Full Product Details