Product Details

SNP ID
rs74072266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:45781539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAATGCGCAGGCCCCAATCCTTT[C/T]TTGCGACTGCAGCATCTGACATGTG
Phenotype
MIM: 615609
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIAH3 PubMed Links

Gene Details

Gene
SIAH3
Gene Name
siah E3 ubiquitin protein ligase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198849.2 2736 UTR 3 NP_942146.2

View Full Product Details