Product Details

SNP ID
rs74041365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23735794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGAGCCTGGAAGGATAATGAAG[A/G]ATAGTTACTGGGGAAGAATCCTGGG
Phenotype
MIM: 602241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIPEP PubMed Links

Gene Details

Gene
MIPEP
Gene Name
mitochondrial intermediate peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005932.3 Intron NP_005923.2
XM_011535097.2 Intron XP_011533399.1
XM_011535098.2 Intron XP_011533400.1

View Full Product Details