Product Details

SNP ID
rs73477126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:76357782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCACAGTGCCAGCCCTTGTGTCC[C/T]CACATTTTTGACACAATAATTTCCT
Phenotype
MIM: 615114
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF516 PubMed Links

Gene Details

Gene
ZNF516
Gene Name
zinc finger protein 516
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014643.3 8526 UTR 3 NP_055458.1
XM_011526269.1 8526 Intron XP_011524571.1
XM_011526270.2 8526 Intron XP_011524572.1
XM_011526271.2 8526 Intron XP_011524573.1
XM_011526272.2 8526 Intron XP_011524574.1
XM_011526273.2 8526 Intron XP_011524575.1
XM_011526274.2 8526 Intron XP_011524576.1
XM_011526275.2 8526 Intron XP_011524577.1
XM_011526276.1 8526 Intron XP_011524578.1
XM_011526277.2 8526 Intron XP_011524579.1
XM_017026097.1 8526 Intron XP_016881586.1

View Full Product Details