Product Details

SNP ID
rs73762693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109297135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACAGCAGCTGGGAGTAAAGGGCG[C/T]CCCCAGGCAAAATCTGCAGGGAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC162P PubMed Links

Gene Details

Gene
CCDC162P
Gene Name
coiled-coil domain containing 162, pseudogene
There are no transcripts associated with this gene.

View Full Product Details