Product Details

SNP ID
rs73766212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:117481250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGAATCAAACCGGGATCCCCTC[C/T]ATTTCTAAAGCCCAAGTTCTTTCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCBLD1 PubMed Links

Gene Details

Gene
DCBLD1
Gene Name
discoidin, CUB and LCCL domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173674.2 Intron NP_775945.1
XM_005266941.1 Intron XP_005266998.1
XM_006715463.1 Intron XP_006715526.1
XM_011535771.1 Intron XP_011534073.1
XM_011535773.2 Intron XP_011534075.1
XM_011535774.2 Intron XP_011534076.1
XM_017010775.1 Intron XP_016866264.1

View Full Product Details