Product Details

SNP ID
rs73766689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:121081330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAATCTGCAATTACTAATGTTGG[G/T]ACTATGCACATCTATACTAATTTGG
Phenotype
MIM: 615867
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TBC1D32 PubMed Links

Gene Details

Gene
TBC1D32
Gene Name
TBC1 domain family member 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152730.5 Intron NP_689943.4
XM_005266861.2 Intron XP_005266918.1
XM_011535569.1 Intron XP_011533871.1
XM_011535570.1 Intron XP_011533872.1
XM_011535571.2 Intron XP_011533873.1
XM_011535574.1 Intron XP_011533876.1
XM_011535575.2 Intron XP_011533877.1
XM_011535576.2 Intron XP_011533878.1
XM_011535580.2 Intron XP_011533882.1
XM_011535582.2 Intron XP_011533884.1
XM_011535585.2 Intron XP_011533887.1
XM_017010397.1 Intron XP_016865886.1
XM_017010398.1 Intron XP_016865887.1
XM_017010399.1 Intron XP_016865888.1
XM_017010400.1 Intron XP_016865889.1
XM_017010401.1 Intron XP_016865890.1
XM_017010402.1 Intron XP_016865891.1
XM_017010403.1 Intron XP_016865892.1
XM_017010404.1 Intron XP_016865893.1
XM_017010405.1 Intron XP_016865894.1

View Full Product Details