Product Details

SNP ID
rs73774524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:134960889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGTCTCCCCAGTACATCTGTG[A/C]CCTTTCTTCTTTAAATCTATGGTTT
Phenotype
MIM: 612450
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HBS1L PubMed Links

Gene Details

Gene
HBS1L
Gene Name
HBS1 like translational GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145158.1 6652 UTR 3 NP_001138630.1
NM_001145207.1 6652 Intron NP_001138679.1
NM_006620.3 6652 UTR 3 NP_006611.1
XM_017010192.1 6652 Intron XP_016865681.1

View Full Product Details