Product Details
- SNP ID
-
rs73596445
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:22972680 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAGTCCTTAGAAGAAGGCCAAGAA[A/G]TAACTCATTAATGAGGTCCAGAGTT
- Phenotype
-
MIM: 609066
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AJUBA
PubMed Links
Gene Details
- Gene
- AJUBA
- Gene Name
- ajuba LIM protein
View Full Product Details