Product Details

SNP ID
rs73307509
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75654152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCTCAGCATAATAAGAGGGGCT[C/T]AATAAATATTATCGCCTCCTGTTTT
Phenotype
MIM: 604576 MIM: 610865 MIM: 612268
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C14orf1 PubMed Links

Gene Details

Gene
C14orf1
Gene Name
chromosome 14 open reading frame 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007176.3 Intron NP_009107.1
Gene
FLVCR2
Gene Name
feline leukemia virus subgroup C cellular receptor family member 2
There are no transcripts associated with this gene.

Gene
TTLL5
Gene Name
tubulin tyrosine ligase like 5
There are no transcripts associated with this gene.

View Full Product Details