Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_194287.2 | 784 | Intron | NP_919263.2 | ||
XM_011536479.1 | 784 | Intron | XP_011534781.1 | ||
XM_011536483.1 | 784 | Intron | XP_011534785.1 | ||
XM_011536487.1 | 784 | Intron | XP_011534789.1 | ||
XM_017021032.1 | 784 | Intron | XP_016876521.1 | ||
XM_017021033.1 | 784 | Intron | XP_016876522.1 | ||
XM_017021034.1 | 784 | Intron | XP_016876523.1 | ||
XM_017021035.1 | 784 | Intron | XP_016876524.1 | ||
XM_017021036.1 | 784 | Intron | XP_016876525.1 | ||
XM_017021037.1 | 784 | UTR 5 | XP_016876526.1 | ||
XM_017021038.1 | 784 | Intron | XP_016876527.1 | ||
XM_017021039.1 | 784 | Intron | XP_016876528.1 | ||
XM_017021040.1 | 784 | Intron | XP_016876529.1 | ||
XM_017021041.1 | 784 | Intron | XP_016876530.1 |