Product Details

SNP ID
rs71413471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77674126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGACACGAGGGACTGCGTGGT[C/T]CAAGAGGCGGCTGAAACCCGACATT
Phenotype
MIM: 605345
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALKBH1 PubMed Links

Gene Details

Gene
ALKBH1
Gene Name
alkB homolog 1, histone H2A dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006020.2 865 Missense Mutation AAC,GAC N286D NP_006011.2

View Full Product Details