Product Details

SNP ID
rs74074348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94018119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGCCCAGTCCAGAACAAGCCCT[C/T]GGTAAATCTGTGTTGTACATCCTCA
Phenotype
MIM: 608338
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00521 PubMed Links

Gene Details

Gene
LINC00521
Gene Name
long intergenic non-protein coding RNA 521
There are no transcripts associated with this gene.

Gene
OTUB2
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023112.3 Intron NP_075601.1
XM_005268055.3 Intron XP_005268112.1
XM_011537148.2 Intron XP_011535450.1

View Full Product Details