Product Details

SNP ID
rs73361323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103561966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACGTTTCAAGCTCTTGGTTTCT[A/G]TCAAACGGCTCGCTCAAGGTGGGTA
Phenotype
MIM: 616003 MIM: 603885
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOPT1 PubMed Links

Gene Details

Gene
APOPT1
Gene Name
apoptogenic 1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302652.1 353 Intron NP_001289581.1
NM_001302653.1 353 Intron NP_001289582.1
NM_001302654.1 353 Intron NP_001289583.1
NM_032374.4 353 Intron NP_115750.2
Gene
BAG5
Gene Name
BCL2 associated athanogene 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015048.2 353 Intron NP_001015048.1
NM_001015049.2 353 Silent Mutation GAC,GAT D14D NP_001015049.1
NM_004873.3 353 Intron NP_004864.1

View Full Product Details