Product Details

SNP ID
rs74091265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105601768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTCAGTGCCAGCCTCAGGCCAG[C/T]GCCCATCAGGCCTGGGGGGCCTGGA
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

View Full Product Details