Product Details
- SNP ID
-
rs73104079
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31303524 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGGGTGATGAAGCATGATTTAAAG[A/G]AACACGCCCTAAGACTATTAAGGGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB116
PubMed Links
Gene Details
- Gene
- DEFB116
- Gene Name
- defensin beta 116
View Full Product Details