Product Details

SNP ID
rs73302664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47735113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTCCCTCTGTCCTGTCCTGCC[C/T]GGTAGGACACACACGCAGCCATTCC
Phenotype
MIM: 606720 MIM: 606057
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENDOU PubMed Links

Gene Details

Gene
ENDOU
Gene Name
endonuclease, poly(U) specific
There are no transcripts associated with this gene.

Gene
LOC105369748
Gene Name
uncharacterized LOC105369748
There are no transcripts associated with this gene.

Gene
RAPGEF3
Gene Name
Rap guanine nucleotide exchange factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098531.2 5540 UTR 3 NP_001092001.1
NM_001098532.2 5540 UTR 3 NP_001092002.1
NM_006105.5 5540 UTR 3 NP_006096.2
XM_005268571.3 5540 Intron XP_005268628.1
XM_011537752.1 5540 Intron XP_011536054.1
XM_011537755.1 5540 Intron XP_011536057.1
XM_011537758.2 5540 Intron XP_011536060.1
XM_017018688.1 5540 Intron XP_016874177.1

View Full Product Details