Product Details

SNP ID
rs73109026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51250982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAAATCGCCCAGGGCTGCAATC[C/T]ATCTTCCTACCATGGATCTAGAGTT
Phenotype
MIM: 607431
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAZAP2 PubMed Links

Gene Details

Gene
DAZAP2
Gene Name
DAZ associated protein 2
There are no transcripts associated with this gene.

Gene
SMAGP
Gene Name
small cell adhesion glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031628.1 Intron NP_001026798.1
NM_001033873.1 Intron NP_001029045.1

View Full Product Details