Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039960.2 | 138 | Intron | NP_001035049.1 | ||
NM_001258401.2 | 138 | Intron | NP_001245330.1 | ||
NM_001258402.1 | 138 | Intron | NP_001245331.1 | ||
NM_001258403.1 | 138 | Intron | NP_001245332.1 | ||
NM_001267615.1 | 138 | Intron | NP_001254544.1 | ||
XM_006719700.1 | 138 | Intron | XP_006719763.1 | ||
XM_011539010.1 | 138 | Intron | XP_011537312.1 | ||
XM_011539011.1 | 138 | Intron | XP_011537313.1 | ||
XM_011539014.2 | 138 | Intron | XP_011537316.1 | ||
XM_017020240.1 | 138 | Intron | XP_016875729.1 | ||
XM_017020241.1 | 138 | UTR 5 | XP_016875730.1 | ||
XM_017020242.1 | 138 | Intron | XP_016875731.1 |