Product Details

SNP ID
rs73103977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53513226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATTCTGGGGTAGGACTTTACCC[C/T]AAAGAGAGACAACACAGCTGATGCA
Phenotype
MIM: 606371 MIM: 604643 MIM: 605053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATF7 PubMed Links

Gene Details

Gene
ATF7
Gene Name
activating transcription factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130060.1 5521 UTR 3 NP_001123532.1
NM_001206682.1 5521 Intron NP_001193611.1
NM_001206683.1 5521 Intron NP_001193612.1
NM_006856.2 5521 UTR 3 NP_006847.1
XM_005268587.3 5521 UTR 3 XP_005268644.1
XM_017018722.1 5521 UTR 3 XP_016874211.1
Gene
LOC100652999
Gene Name
uncharacterized LOC100652999
There are no transcripts associated with this gene.

Gene
NPFF
Gene Name
neuropeptide FF-amide peptide precursor
There are no transcripts associated with this gene.

Gene
TARBP2
Gene Name
TARBP2, RISC loading complex RNA binding subunit
There are no transcripts associated with this gene.

View Full Product Details