Product Details

SNP ID
rs74097651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:63142785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAACTTATTTTTGTTTATAGAA[A/G]CAAGACAACACACAGTAAAATTGAG
Phenotype
MIM: 600821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AVPR1A PubMed Links

Gene Details

Gene
AVPR1A
Gene Name
arginine vasopressin receptor 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000706.4 7805 UTR 3 NP_000697.1

View Full Product Details