Product Details

SNP ID
rs67133230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:45730630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACATCGATAATCGGCTGCGAAAT[A/G]ATCCCGGCAGCCGGGGGCACAGCCT
Phenotype
MIM: 609539
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARID2 PubMed Links

Gene Details

Gene
ARID2
Gene Name
AT-rich interaction domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152641.2 Intron NP_689854.2
XM_006719272.3 Intron XP_006719335.1
XM_011538025.1 Intron XP_011536327.1
Gene
LINC00938
Gene Name
long intergenic non-protein coding RNA 938
There are no transcripts associated with this gene.

View Full Product Details