Product Details

SNP ID
rs74149212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91424804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAACATAAACTCTAAAGGGTTAT[A/C]TTTTTACCTGAACTTGTAAAGTTGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HECTD2 PubMed Links

Gene Details

Gene
HECTD2
Gene Name
HECT domain E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284274.1 Intron NP_001271203.1
NM_173497.3 Intron NP_775768.3
NM_182765.4 Intron NP_877497.3
XM_017015750.1 Intron XP_016871239.1
XM_017015751.1 Intron XP_016871240.1
XM_017015752.1 Intron XP_016871241.1
Gene
HECTD2-AS1
Gene Name
HECTD2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details