Product Details

SNP ID
rs72813685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95988363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGGTAACAACACAGTATTATCA[A/G]TATATTTGCATTTGTTTTTGTGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CC2D2B PubMed Links

Gene Details

Gene
CC2D2B
Gene Name
coiled-coil and C2 domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001732.3 Intron NP_001001732.2
NM_001159747.1 Intron NP_001153219.1
XM_011539789.2 Intron XP_011538091.1
XM_011539790.2 Intron XP_011538092.1
XM_011539791.2 Intron XP_011538093.1
XM_011539792.2 Intron XP_011538094.1
XM_011539793.2 Intron XP_011538095.1
XM_011539795.2 Intron XP_011538097.1
XM_017016240.1 Intron XP_016871729.1
Gene
ENTPD1-AS1
Gene Name
ENTPD1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details