Product Details

SNP ID
rs74158816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116672551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTACATAGTCCATAGCTTGGCATC[A/G]GCACAGATCGATGCTTAGTGAGCAC
Phenotype
MIM: 610701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf82 PubMed Links

Gene Details

Gene
C10orf82
Gene Name
chromosome 10 open reading frame 82
There are no transcripts associated with this gene.

Gene
HSPA12A
Gene Name
heat shock protein family A (Hsp70) member 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025015.2 4085 UTR 3 NP_079291.2
XM_005269672.3 4085 UTR 3 XP_005269729.1
XM_005269673.4 4085 UTR 3 XP_005269730.1
XM_011539579.2 4085 UTR 3 XP_011537881.1
XM_011539580.2 4085 UTR 3 XP_011537882.1
XM_017016032.1 4085 UTR 3 XP_016871521.1
Gene
LOC105378499
Gene Name
uncharacterized LOC105378499
There are no transcripts associated with this gene.

View Full Product Details