Product Details

SNP ID
rs73213893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:14088102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCATCTTTTCTCCTCTTATGA[C/T]ATATAATTTAAAATTTCATTTTTCT
Phenotype
MIM: 608113
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGCZ PubMed Links

Gene Details

Gene
SGCZ
Gene Name
sarcoglycan zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322879.1 3940 UTR 3 NP_001309808.1
NM_001322880.1 3940 UTR 3 NP_001309809.1
NM_001322881.1 3940 UTR 3 NP_001309810.1
NM_139167.3 3940 UTR 3 NP_631906.2

View Full Product Details