Product Details
- SNP ID
-
rs73204871
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:12948999 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAACCAGAAAAACACGTGTTCA[A/G]CGTTTCATAGTTGTCTAATTTCTGT
- Phenotype
-
MIM: 615666
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1456
PubMed Links
Gene Details
- Gene
- KIAA1456
- Gene Name
- KIAA1456
View Full Product Details