Product Details

SNP ID
rs72964451
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:43148849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCTCCACACACTTAAAAAAAG[A/G]CACAAGTCCTGTTTATTAACATTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM183A PubMed Links

Gene Details

Gene
FAM183A
Gene Name
family with sequence similarity 183 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101376.2 Intron NP_001094846.2
XM_005270875.4 Intron XP_005270932.1
XM_005270876.4 Intron XP_005270933.1
XM_017001322.1 Intron XP_016856811.1

View Full Product Details