Product Details

SNP ID
rs72896537
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:47217856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTTAGACAGATGGGGCTCCTCA[C/T]AGGATCTTCCAAACGTTGGAAAGGA
Phenotype
MIM: 187040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAL1 PubMed Links

Gene Details

Gene
TAL1
Gene Name
TAL bHLH transcription factor 1, erythroid differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287347.2 3186 UTR 3 NP_001274276.1
NM_001290403.1 3186 UTR 3 NP_001277332.1
NM_001290404.1 3186 UTR 3 NP_001277333.1
NM_001290405.1 3186 UTR 3 NP_001277334.1
NM_001290406.1 3186 UTR 3 NP_001277335.1
NM_003189.5 3186 UTR 3 NP_003180.1
XM_005271160.4 3186 UTR 3 XP_005271217.1
XM_017002187.1 3186 UTR 3 XP_016857676.1
XM_017002188.1 3186 UTR 3 XP_016857677.1
XM_017002189.1 3186 UTR 3 XP_016857678.1
XM_017002190.1 3186 UTR 3 XP_016857679.1
XM_017002191.1 3186 UTR 3 XP_016857680.1
XM_017002192.1 3186 Intron XP_016857681.1
XM_017002193.1 3186 UTR 3 XP_016857682.1

View Full Product Details