Product Details

SNP ID
rs72904275
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:51287620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTGGGTGTCTGTGTACATCCTC[A/G]TGCCCAACCCCAGAAGCATTGAAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC39A PubMed Links

Gene Details

Gene
TTC39A
Gene Name
tetratricopeptide repeat domain 39A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080494.3 2478 UTR 3 NP_001073963.1
NM_001144832.2 2478 UTR 3 NP_001138304.1
NM_001297662.1 2478 Intron NP_001284591.1
NM_001297663.1 2478 UTR 3 NP_001284592.1
NM_001297664.1 2478 UTR 3 NP_001284593.1
NM_001297665.1 2478 UTR 3 NP_001284594.1
NM_001297666.1 2478 UTR 3 NP_001284595.1
NM_001297667.1 2478 UTR 3 NP_001284596.1
XM_005270644.2 2478 UTR 3 XP_005270701.1
XM_006710471.2 2478 UTR 3 XP_006710534.1
XM_011541048.2 2478 UTR 3 XP_011539350.1
XM_011541049.2 2478 UTR 3 XP_011539351.1
XM_011541050.2 2478 UTR 3 XP_011539352.1
XM_011541051.2 2478 UTR 3 XP_011539353.1
XM_011541052.2 2478 UTR 3 XP_011539354.1
XM_011541053.2 2478 UTR 3 XP_011539355.1
XM_011541054.2 2478 UTR 3 XP_011539356.1
XM_017000715.1 2478 UTR 3 XP_016856204.1

View Full Product Details